Assay Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277672.1 | NP_001264601.1 | 2 - 3 | 650 | 73 | |
| GenBank mRNA | BX929763.1 | - | 2 - 3 | 647 | 73 | |
| BX950281.2 | - | 2 - 3 | 650 | 73 | ||
More Information
Set Membership: |
|

RAB32
Genomic Map