Assay Details
| Gene Symbol: TTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944815.3 | XP_004944872.1 | 3 - 4 | 466 | 79 | |
| XM_414484.6 | XP_414484.3 | 3 - 4 | 448 | 79 | ||
| GenBank mRNA | BX929863.1 | - | 2 - 3 | 419 | 79 | |
More Information
Set Membership: |
|

TTC1
Genomic Map