Assay Details
| Gene Symbol: SERHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277373.1 | NP_001264302.1 | 3 - 4 | 414 | 96 | |
| XM_015285070.2 | XP_015140556.1 | 3 - 4 | 378 | 96 | ||
| XM_025148464.1 | XP_025004232.1 | 3 - 4 | 525 | 96 | ||
| GenBank mRNA | BX929989.1 | - | 2 - 3 | 321 | 96 | |
| BX932388.1 | - | 2 - 3 | 301 | 96 | ||
| CD218551.1 | - | 3 - 4 | 383 | 96 | ||
More Information
Set Membership: |
|

SERHL
Genomic Map