Assay Details
Gene Symbol: H2AFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277338.2 | NP_001264267.1 | 2 - 3 | 344 | 67 | |
XM_015288210.1 | XP_015143696.1 | 3 - 4 | 843 | 67 | ||
XM_015288211.2 | XP_015143697.1 | 3 - 4 | 489 | 67 | ||
GenBank mRNA | BX930607.1 | - | 3 - 4 | 463 | 67 | |
BX931423.2 | - | 3 - 4 | 390 | 67 |
Gene Symbol: LOC112532616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151724.1 | XP_025007492.1 | 3 - 4 | 487 | 67 | |
XM_025151725.1 | XP_025007493.1 | 3 - 4 | 495 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
