Assay Details
| Gene Symbol: SLC35E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418259.6 | XP_418259.3 | 2 - 3 | 527 | 79 | |
| GenBank mRNA | BX930649.1 | - | 2 - 3 | 516 | 79 | |
More Information
Set Membership: |
|

SLC35E1
Genomic Map