Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256506.1 | NP_001243435.1 | 1 - 2 | 60 | 69 | |
| NM_001256507.1 | NP_001243436.1 | 1 - 2 | 60 | 69 | ||
| GenBank mRNA | BX930770.1 | - | 1 - 2 | 46 | 69 | |
| BX931155.1 | - | 1 - 2 | 50 | 69 | ||
| BX933521.2 | - | 1 - 2 | 60 | 69 | ||
| BX934114.1 | - | 1 - 2 | 50 | 69 | ||
| BX935247.2 | - | 1 - 2 | 52 | 69 | ||
More Information
Set Membership: |
|

BORCS8
Genomic Map