Assay Details
| Gene Symbol: AKAP17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195550.1 | NP_001182479.1 | 1 - 2 | 77 | 69 | |
| GenBank mRNA | BX931250.1 | - | 1 - 2 | 92 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195550.1 transcripts.
Set Membership: |
|

AKAP17A
Genomic Map