Assay Details
| Gene Symbol: AKAP17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195550.1 | NP_001182479.1 | 2 - 3 | 1282 | 108 | |
| XM_025149890.1 | XP_025005658.1 | 1 - 2 | 1210 | 108 | ||
| XM_025149894.1 | XP_025005662.1 | 2 - 3 | 974 | 108 | ||
| GenBank mRNA | BX931250.1 | - | 3 - 4 | 1059 | 108 | |
| CR354007.1 | - | 2 - 3 | 1282 | 108 | ||
More Information
Set Membership: |
|

AKAP17A
Genomic Map