Assay Details
| Gene Symbol: SLC25A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414400.6 | XP_414400.1 | 2 - 3 | 270 | 67 | |
| GenBank mRNA | BX931997.1 | - | 2 - 3 | 251 | 67 | |
| BX932308.2 | - | 2 - 3 | 238 | 67 | ||
More Information
Set Membership: |
|

SLC25A20
Genomic Map