Assay Details
| Gene Symbol: ACP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278076.1 | NP_001265005.1 | 5 - 6 | 626 | 55 | |
| NM_001278077.1 | NP_001265006.1 | 4 - 5 | 546 | 55 | ||
| XM_015296397.2 | XP_015151883.2 | 5 - 6 | 6049 | 55 | ||
| GenBank mRNA | BU318938.1 | - | 5 - 6 | 480 | 55 | |
| BU324326.1 | - | 3 - 4 | 358 | 55 | ||
| BX932077.1 | - | 5 - 6 | 626 | 55 | ||
| BX932300.1 | - | 3 - 4 | 408 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278077.1 transcripts.
Set Membership: |
|

ACP6
Genomic Map