Assay Details
| Gene Symbol: TAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277561.1 | NP_001264490.1 | 9 - 10 | 1300 | 66 | |
| GenBank mRNA | BX929859.2 | - | 9 - 10 | 1278 | 66 | |
| BX932342.1 | - | 7 - 8 | 881 | 66 | ||
More Information
Set Membership: |
|

TAF1A
Genomic Map