Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419549.6 | XP_419549.2 | 1 - 2 | 291 | 104 | |
| GenBank mRNA | BX930693.2 | - | 1 - 2 | 58 | 104 | |
| BX932346.1 | - | 1 - 2 | 296 | 104 | ||
More Information
Set Membership: |
|

MAP1LC3C
Genomic Map