Assay Details
| Gene Symbol: RPL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257333.2 | NP_001244262.1 | 7 - 8 | 642 | 80 | |
| GenBank mRNA | BU409272.1 | - | 6 - 7 | 589 | 80 | |
| BX929503.1 | - | 6 - 7 | 592 | 80 | ||
| BX932602.1 | - | 5 - 6 | 668 | 80 | ||
| BX933063.1 | - | 7 - 8 | 634 | 80 | ||
| BX933942.2 | - | 6 - 7 | 755 | 80 | ||
| BX934426.1 | - | 6 - 7 | 610 | 80 | ||
| CR338955.1 | - | 6 - 7 | 607 | 80 | ||
More Information
Set Membership: |
|

RPL9
Genomic Map