Assay Details
| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302135.1 | NP_001289064.1 | 1 - 2 | 249 | 120 | |
| GenBank mRNA | BX931073.1 | - | 1 - 2 | 175 | 120 | |
| BX933546.1 | - | 1 - 2 | 177 | 120 | ||
| CO635380.1 | - | 1 - 2 | 249 | 120 | ||
More Information
Set Membership: |
|

GNG5
Genomic Map