Assay Details
| Gene Symbol: RPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277164.1 | NP_001264093.1 | 6 - 7 | 712 | 79 | |
| GenBank mRNA | BX933609.1 | - | 5 - 6 | 698 | 79 | |
| BX934597.2 | - | 5 - 6 | 698 | 79 | ||
| BX935640.2 | - | 5 - 6 | 698 | 79 | ||
More Information
Set Membership: |
|

RPS2
Genomic Map