Assay Details
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416953.6 | XP_416953.2 | 1 - 2 | 284 | 150 | |
| GenBank mRNA | BX930435.2 | - | 1 - 2 | 274 | 150 | |
| BX930702.2 | - | 1 - 2 | 259 | 150 | ||
| BX933992.1 | - | 1 - 2 | 257 | 150 | ||
More Information
Set Membership: |
|

RAB20
Genomic Map