Assay Details
| Gene Symbol: LACTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277407.1 | NP_001264336.1 | 6 - 7 | 889 | 125 | |
| XM_025147086.1 | XP_025002854.1 | 6 - 7 | 897 | 124 | ||
| GenBank mRNA | BX929443.1 | - | 6 - 7 | 844 | 125 | |
| BX935889.1 | - | 6 - 7 | 845 | 125 | ||
More Information
Set Membership: |
|

LACTB2
Genomic Map