Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142137.1 | XP_024997905.1 | 1 - 2 | 176 | 101 | |
| GenBank mRNA | BX950729.1 | - | 3 - 4 | 549 | 101 | |
More Information
Set Membership: |
|

TMEM248
Genomic Map