Assay Details
| Gene Symbol: PPP1R1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289551.2 | XP_015145037.1 | 1 - 2 | 292 | 70 | |
| XM_421970.6 | XP_421970.3 | 1 - 2 | 346 | 70 | ||
| GenBank mRNA | BX929596.2 | - | 1 - 2 | 312 | 70 | |
| BX929642.2 | - | 1 - 2 | 312 | 70 | ||
More Information
Set Membership: |
|

PPP1R1C
Genomic Map