Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939794.3 | XP_004939851.1 | 9 - 10 | 780 | 70 | |
| XM_419166.6 | XP_419166.3 | 9 - 10 | 765 | 70 | ||
| GenBank mRNA | BX930334.1 | - | 9 - 10 | 749 | 70 | |
| BX930593.2 | - | 9 - 10 | 747 | 70 | ||
| BX931215.1 | - | 9 - 10 | 749 | 70 | ||
| BX934058.2 | - | 8 - 9 | 682 | 70 | ||
More Information
Set Membership: |
|

IMPACT
Genomic Map