Assay Details
| Gene Symbol: EXOSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252154.1 | NP_001239083.1 | 6 - 7 | 67 | ||
| GenBank mRNA | BX932890.2 | - | 6 - 7 | 638 | 67 | |
| BX933850.1 | - | 6 - 7 | 613 | 67 | ||
| BX935756.1 | - | 6 - 7 | 67 | |||
More Information
Set Membership: |
|

EXOSC7
Genomic Map