Assay Details
| Gene Symbol: SOCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294233.2 | XP_015149719.1 | 2 | 830 | 155 | |
| GenBank mRNA | BX933215.2 | - | 1 | 656 | 155 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOCS1| Gene Symbol: SOCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294233.2 | XP_015149719.1 | 2 | 830 | 155 | |
| GenBank mRNA | BX933215.2 | - | 1 | 656 | 155 | |
Set Membership: |
|