Assay Details
| Gene Symbol: GPR158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257299.1 | NP_001244228.1 | 7 - 8 | 1695 | 66 | |
| GenBank mRNA | BX934637.2 | - | 3 - 4 | 415 | 66 | |
More Information
Set Membership: |
|

GPR158
Genomic Map