Assay Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277682.1 | NP_001264611.1 | 1 - 2 | 115 | 83 | |
| XM_015279128.2 | XP_015134614.1 | 2 - 3 | 203 | 83 | ||
| GenBank mRNA | BX935768.2 | - | 1 - 2 | 115 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277682.1, XM_015279128.2 transcripts.
Set Membership: |
|

PSKH1
Genomic Map