Assay Details
| Gene Symbol: AAGAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271528.1 | NP_001258457.1 | 1 - 2 | 92 | 54 | |
| XM_015292097.2 | XP_015147583.1 | 1 - 2 | 131 | 54 | ||
| GenBank mRNA | BX929773.2 | - | 1 - 2 | 69 | 54 | |
| BX930322.2 | - | 1 - 2 | 69 | 54 | ||
| BX935916.2 | - | 1 - 2 | 93 | 54 | ||
More Information
Set Membership: |
|

AAGAB
Genomic Map