Assay Details
| Gene Symbol: NSMCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414884.6 | XP_414884.1 | 2 - 3 | 342 | 87 | |
| GenBank mRNA | BX936090.2 | - | 2 - 3 | 324 | 87 | |
More Information
Set Membership: |
|

NSMCE1
Genomic Map