Assay Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277821.1 | NP_001264750.1 | 3 - 4 | 416 | 72 | |
| GenBank mRNA | BU127447.1 | - | 3 - 4 | 393 | 72 | |
| BX929471.1 | - | 2 - 3 | 242 | 72 | ||
| BX932299.2 | - | 3 - 4 | 359 | 72 | ||
| BX932961.2 | - | 3 - 4 | 364 | 72 | ||
| CR353386.1 | - | 3 - 4 | 416 | 72 | ||
| CR385451.1 | - | 3 - 4 | 409 | 72 | ||
| CR406248.1 | - | 3 - 4 | 404 | 72 | ||
More Information
Set Membership: |
|

COMMD8
Genomic Map