Assay Details
| Gene Symbol: ATP5C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278096.1 | NP_001265025.1 | 7 - 8 | 815 | 117 | |
| NM_001278097.1 | NP_001265026.1 | 7 - 8 | 815 | 117 | ||
| XM_015293766.2 | XP_015149252.1 | 7 - 8 | 855 | 117 | ||
| GenBank mRNA | BX936136.1 | - | 7 - 8 | 802 | 117 | |
| BX950663.1 | - | 7 - 8 | 783 | 117 | ||
| CR353583.1 | - | 7 - 8 | 815 | 117 | ||
More Information
Set Membership: |
|

ATP5C1
Genomic Map