Assay Details
| Gene Symbol: NARFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277555.1 | NP_001264484.1 | 5 - 6 | 583 | 64 | |
| XM_015294191.2 | XP_015149677.1 | 5 - 6 | 64 | |||
| GenBank mRNA | BX934839.1 | - | 5 - 6 | 583 | 64 | |
| CR385778.1 | - | 5 - 6 | 583 | 64 | ||
More Information
Set Membership: |
|

NARFL
Genomic Map