Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422056.6 | XP_422056.3 | 4 - 5 | 844 | 60 | |
| GenBank mRNA | CR387538.1 | - | 2 - 3 | 413 | 60 | |
More Information
Set Membership: |
|

CYP27A1
Genomic Map