Assay Details
| Gene Symbol: SGK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946998.2 | XP_004947055.1 | 15 | 2199 | 103 | |
| XM_015296774.2 | XP_015152260.2 | 14 | 2136 | 103 | ||
| XM_015296775.2 | XP_015152261.1 | 15 | 2199 | 103 | ||
| XM_015296776.1 | XP_015152262.1 | 16 | 2280 | 103 | ||
| XM_015296777.2 | XP_015152263.1 | 14 | 3051 | 103 | ||
| XM_015296778.2 | XP_015152264.1 | 14 | 2135 | 103 | ||
| XM_015296779.2 | XP_015152265.1 | 14 | 2240 | 103 | ||
| XM_015296780.2 | XP_015152266.1 | 14 | 2143 | 103 | ||
| XM_015296781.1 | XP_015152267.1 | 14 | 2040 | 103 | ||
| XM_025142303.1 | XP_024998071.1 | 16 | 2279 | 103 | ||
| GenBank mRNA | CR387909.1 | - | 1 | 850 | 103 | |
More Information
Set Membership: |
|

SGK2
Genomic Map