Assay Details
| Gene Symbol: RHOBTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232709.5 | XP_001232710.3 | 3 - 4 | 425 | 77 | |
| XM_015297318.2 | XP_015152804.1 | 3 - 4 | 408 | 77 | ||
| XM_015297319.2 | XP_015152805.1 | 2 - 3 | 475 | 77 | ||
| GenBank mRNA | AJ851586.1 | - | 2 - 3 | 477 | 77 | |
More Information
Set Membership: |
|

RHOBTB2
Genomic Map