Assay Details
| Gene Symbol: CYB561A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199574.1 | NP_001186503.1 | 4 - 5 | 698 | 110 | |
| XM_015286081.2 | XP_015141567.1 | 4 - 5 | 743 | 110 | ||
| XM_015286082.2 | XP_015141568.1 | 4 - 5 | 608 | 110 | ||
| XM_015286083.2 | XP_015141569.1 | 4 - 5 | 688 | 110 | ||
| GenBank mRNA | BX930104.2 | - | 3 - 4 | 588 | 110 | |
| BX934774.2 | - | 4 - 5 | 648 | 110 | ||
| BX950333.1 | - | 4 - 5 | 698 | 110 | ||
| DN928923.1 | - | 1 - 2 | 113 | 110 | ||
More Information
Set Membership: |
|

CYB561A3
Genomic Map