Assay Details
| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302111.1 | NP_001289040.1 | 1 - 2 | 151 | 73 | |
| GenBank mRNA | BX930263.2 | - | 1 - 2 | 151 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302111.1 transcripts.
Set Membership: |
|

SPTSSB
Genomic Map