Assay Details
| Gene Symbol: SLC35E2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR386029.1 | - | 4 - 5 | 486 | 73 | |
| CR406890.1 | - | 2 - 3 | 350 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC35E2B| Gene Symbol: SLC35E2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR386029.1 | - | 4 - 5 | 486 | 73 | |
| CR406890.1 | - | 2 - 3 | 350 | 73 | ||
Set Membership: |
|