Assay Details
| Gene Symbol: KIAA0391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421241.5 | XP_421241.2 | 5 - 6 | 1861 | 59 | |
| GenBank mRNA | CR389599.1 | - | 3 - 4 | 278 | 59 | |
More Information
Set Membership: |
|

KIAA0391
Genomic Map