Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199250.1 | NP_001186179.1 | 4 - 5 | 377 | 59 | |
| XM_025148546.1 | XP_025004314.1 | 4 - 5 | 450 | 59 | ||
| GenBank mRNA | CR390140.1 | - | 4 - 5 | 303 | 59 | |
More Information
Set Membership: |
|

RBX1
Genomic Map