Assay Details
| Gene Symbol: PGR2/3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001329217.1 | NP_001316146.1 | 4 - 5 | 390 | 102 | |
| GenBank mRNA | BU472863.1 | - | 4 - 5 | 393 | 102 | |
| CR390223.1 | - | 4 - 5 | 406 | 102 | ||
More Information
Set Membership: |
|

PGR2/3
Genomic Map