Assay Details
| Gene Symbol: SEC61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302159.1 | NP_001289088.1 | 2 - 3 | 166 | 138 | |
| GenBank mRNA | CR390434.1 | - | 2 - 3 | 167 | 138 | |
More Information
Set Membership: |
|

SEC61B
Genomic Map