Assay Details
| Gene Symbol: PRR14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015275419.2 | XP_015130905.1 | 5 - 6 | 6410 | 68 | |
| XM_415250.6 | XP_415250.4 | 5 - 6 | 6188 | 68 | ||
| GenBank mRNA | CR390992.1 | - | 2 - 3 | 475 | 68 | |
More Information
Set Membership: |
|

PRR14L
Genomic Map