Assay Details
| Gene Symbol: MED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938702.3 | XP_004938759.1 | 1 - 2 | 164 | 83 | |
| GenBank mRNA | BX930642.1 | - | 1 - 2 | 39 | 83 | |
| CR407285.1 | - | 1 - 2 | 169 | 83 | ||
More Information
Set Membership: |
|

MED4
Genomic Map