Assay Details
| Gene Symbol: GABRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167703.1 | NP_001161175.1 | 5 - 6 | 761 | 64 | |
| NM_001167704.1 | NP_001161176.1 | 5 - 6 | 761 | 64 | ||
| GenBank mRNA | X56647.1 | - | 761 | 64 | ||
| X56648.1 | - | 464 | 64 | |||
More Information
Set Membership: |
|

GABRQ
Genomic Map