Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198600.1 | NP_001185529.1 | 3 - 4 | 1171 | 126 | |
| XM_015275703.2 | XP_015131189.1 | 4 - 5 | 1286 | 126 | ||
| GenBank mRNA | AJ295847.1 | - | 3 - 4 | 1171 | 126 | |
| BX931005.1 | - | 3 - 4 | 1087 | 126 | ||
More Information
Set Membership: |
|

LRRC17
Genomic Map