Assay Details
| Gene Symbol: TMEM208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256963.1 | NP_001243892.1 | 5 - 6 | 401 | 72 | |
| XM_015279105.2 | XP_015134591.1 | 4 - 5 | 303 | 72 | ||
| GenBank mRNA | AJ720252.1 | - | 5 - 6 | 443 | 72 | |
| BU296314.1 | - | 5 - 6 | 401 | 72 | ||
More Information
Set Membership: |
|

TMEM208
Genomic Map