Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935430.3 | XP_004935487.1 | 6 - 7 | 1066 | 74 | |
| XM_015283744.2 | XP_015139230.1 | 5 - 6 | 912 | 74 | ||
| GenBank mRNA | CR388477.1 | - | 3 - 4 | 319 | 74 | |
More Information
Set Membership: |
|

CNST
Genomic Map