Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 7 - 8 | 673 | 151 | |
| XM_025155367.1 | XP_025011135.1 | 8 - 9 | 860 | 151 | ||
| XM_025155368.1 | XP_025011136.1 | 8 - 9 | 772 | 151 | ||
| XM_025155369.1 | XP_025011137.1 | 5 - 6 | 564 | 151 | ||
| GenBank mRNA | CR390555.1 | - | 2 - 3 | 185 | 151 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map