Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 9 - 10 | 808 | 137 | |
| XM_025155367.1 | XP_025011135.1 | 10 - 11 | 995 | 137 | ||
| XM_025155368.1 | XP_025011136.1 | 10 - 11 | 907 | 137 | ||
| XM_025155369.1 | XP_025011137.1 | 7 - 8 | 699 | 137 | ||
| GenBank mRNA | CR390555.1 | - | 4 - 5 | 320 | 137 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map