Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 10 - 11 | 913 | 96 | |
| XM_025155367.1 | XP_025011135.1 | 11 - 12 | 1100 | 96 | ||
| XM_025155368.1 | XP_025011136.1 | 11 - 12 | 1012 | 96 | ||
| XM_025155369.1 | XP_025011137.1 | 8 - 9 | 804 | 96 | ||
| GenBank mRNA | CR390555.1 | - | 5 - 6 | 425 | 96 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map