Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 11 - 12 | 962 | 76 | |
| XM_025155367.1 | XP_025011135.1 | 12 - 13 | 1149 | 76 | ||
| XM_025155368.1 | XP_025011136.1 | 12 - 13 | 1061 | 76 | ||
| XM_025155369.1 | XP_025011137.1 | 9 - 10 | 853 | 76 | ||
| GenBank mRNA | CR390555.1 | - | 6 - 7 | 474 | 76 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map