Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 13 - 14 | 1138 | 137 | |
| XM_025155367.1 | XP_025011135.1 | 14 - 15 | 1325 | 137 | ||
| XM_025155368.1 | XP_025011136.1 | 14 - 15 | 1237 | 137 | ||
| XM_025155369.1 | XP_025011137.1 | 11 - 12 | 1029 | 137 | ||
| GenBank mRNA | CR390555.1 | - | 8 - 9 | 650 | 137 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map